Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042594.1 | NP_001036059.1 | 9 | 1208 | ||
NM_015450.2 | NP_056265.2 | 10 | 1339 | |||
NR_003102.1 | 10 | 1339 | ||||
NR_003103.1 | 10 | 1339 | ||||
NR_003104.1 | 10 | 1339 | ||||
XM_006715917.3 | XP_006715980.1 | 10 | 1329 | |||
XM_011516006.1 | XP_011514308.1 | 11 | 1451 | |||
XM_011516007.1 | XP_011514309.1 | 11 | 1450 | |||
XM_017011941.1 | XP_016867430.1 | 10 | 1337 | |||
XM_017011942.1 | XP_016867431.1 | 9 | 1199 | |||
GenBank mRNA | AK001230.1 | - | 6 | 761 | ||
AK001935.1 | - | 7 | 868 | |||
AK022580.1 | - | 10 | 1176 | |||
AK302263.1 | - | 10 | 1494 | |||
AY823520.1 | - | 6 | 749 | |||
AY823521.1 | - | 6 | 749 | |||
AY823522.1 | - | 6 | 749 | |||
AY823523.1 | - | 5 | 618 | |||
AY823524.1 | - | 6 | 749 | |||
BC002923.2 | - | 7 | 795 | 3955573 |