Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318520.1 | NP_001305449.1 | 9 | 1099 | ||
NM_173630.3 | NP_775901.3 | 9 | 1099 | |||
XM_006722434.3 | XP_006722497.1 | 9 | 1099 | |||
XM_006722435.3 | XP_006722498.1 | 9 | 1099 | |||
XM_011525902.2 | XP_011524204.1 | 9 | 1099 | |||
XM_011525903.2 | XP_011524205.1 | 9 | 1099 | |||
XM_011525904.2 | XP_011524206.1 | 9 | 1099 | |||
XM_011525905.2 | XP_011524207.1 | 9 | 1099 | |||
XM_011525907.2 | XP_011524209.1 | 9 | 1099 | |||
XM_011525908.2 | XP_011524210.1 | 9 | 1099 | |||
XM_017025693.1 | XP_016881182.1 | 9 | 1099 | |||
XM_017025694.1 | XP_016881183.1 | 6 | 654 | |||
XM_017025695.1 | XP_016881184.1 | 2 | 156 | |||
XM_017025696.1 | XP_016881185.1 | 9 | 1034 | |||
GenBank mRNA | AK096404.1 | - | 9 | 1105 | ||
BC143723.1 | - | 9 | 1076 | |||
CR749799.1 | - | 4 | 404 |