Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THUMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114092.1 | NP_001107564.1 | 3 | 453 | ||
NM_015453.2 | NP_056268.2 | 3 | 658 | |||
XM_005265021.4 | XP_005265078.1 | 3 | 1015 | |||
XM_005265022.4 | XP_005265079.1 | 3 | 1218 | |||
XM_005265023.3 | XP_005265080.1 | 3 | 601 | |||
XM_005265024.4 | XP_005265081.1 | 3 | 601 | |||
XM_006713086.3 | XP_006713149.1 | 3 | 1257 | |||
XM_011533570.2 | XP_011531872.1 | 3 | 721 | |||
XM_011533571.2 | XP_011531873.1 | 3 | 658 | |||
XM_017006113.1 | XP_016861602.1 | 3 | 1015 | |||
XM_017006114.1 | XP_016861603.1 | 3 | 1257 | |||
XM_017006115.1 | XP_016861604.1 | 3 | 658 | |||
XM_017006116.1 | XP_016861605.1 | 3 | 644 | |||
XM_017006117.1 | XP_016861606.1 | 3 | 1015 | |||
XM_017006118.1 | XP_016861607.1 | 3 | 1257 | |||
XM_017006119.1 | XP_016861608.1 | 3 | 644 | |||
GenBank mRNA | AK001680.1 | - | 3 | 417 | ||
AK023263.1 | - | 3 | 639 | |||
BC001622.1 | - | 3 | 432 | 3355978 | ||
BC010421.1 | - | 3 | 989 | 3140059 |