Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000155.3 | NP_000146.2 | 10 | 1123 | ||
NM_001258332.1 | NP_001245261.1 | 8 | 998 | |||
GenBank mRNA | AK096026.1 | - | 5 | 1415 | ||
AK096137.1 | - | 7 | 1795 | |||
AK314340.1 | - | 10 | 1105 | |||
BC015045.2 | - | 10 | 1048 | 3922902 | ||
BT009852.1 | - | 10 | 1007 | |||
KU178089.1 | - | 10 | 1007 | |||
KU178090.1 | - | 10 | 1074 | |||
M60091.1 | - | 10 | 1035 |