Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015473.3 | NP_056288.2 | 30 | 4929 | ||
GenBank mRNA | AB037737.1 | - | 21 | 3377 | ||
AK000343.1 | - | 8 | 1215 | |||
AK023789.1 | - | 12 | 1936 | |||
BC045764.1 | - | 24 | 3942 | |||
BC063867.1 | - | 12 | 1953 | |||
CR749444.1 | - | 23 | 3627 |