Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000156.5 | NP_000147.1 | 2 | 314 | ||
NM_138924.2 | NP_620279.1 | 2 | 314 | |||
GenBank mRNA | AK289465.1 | - | 2 | 288 | ||
AK297487.1 | - | 2 | 288 | |||
BC016760.2 | - | 2 | 261 | 2989341 | ||
BC017936.1 | - | 2 | 259 | 4246521 | ||
BI914772.1 | - | 2 | 297 | |||
BT007034.1 | - | 2 | 220 | |||
EU794612.1 | - | 2 | 220 | |||
Z49878.1 | - | 2 | 263 |