Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIN3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145357.1 | NP_001138829.1 | 11 | 1824 | ||
NM_001145358.1 | NP_001138830.1 | 11 | 1999 | |||
NM_015477.2 | NP_056292.1 | 11 | 1901 | |||
XM_006720465.3 | XP_006720528.1 | 11 | 1707 | |||
XM_006720466.3 | XP_006720529.1 | 12 | 2097 | |||
XM_006720467.3 | XP_006720530.1 | 13 | 1915 | |||
GenBank mRNA | AF418569.1 | - | 11 | 1901 | ||
AK027559.1 | - | 11 | 1901 | |||
AK096477.1 | - | 11 | 1950 | |||
AL832463.1 | - | 11 | 1825 | |||
AY044430.1 | - | 10 | 1687 | |||
BC137098.1 | - | 11 | 1752 | 9021095 | ||
BC137099.1 | - | 11 | 1752 | 9021096 | ||
BX649162.1 | - | 11 | 1938 |