Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136191.2 | NP_001129663.1 | 10 | 2541 | ||
NM_015493.6 | NP_056308.3 | 8 | 2283 | |||
GenBank mRNA | AB040951.2 | - | 9 | 2493 | ||
AB284125.1 | - | 11 | 2724 | |||
AK002094.1 | - | 7 | 973 | |||
AK023332.1 | - | 7 | 1025 | |||
AY639929.1 | - | 8 | 2204 | |||
BC082762.1 | - | 10 | 2495 | 6497026 | ||
BC098105.1 | - | 9 | 2293 | 40012030 | ||
BC098286.1 | - | 9 | 2293 | 40012033 | ||
BC098312.1 | - | 9 | 2293 | 40012032 |