Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM87A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110503.2 | NP_001103973.1 | 6 | 663 | ||
NM_001286487.1 | NP_001273416.1 | 5 | 602 | |||
NM_015497.4 | NP_056312.2 | 6 | 663 | |||
XM_005254287.1 | XP_005254344.1 | 6 | 514 | |||
XM_006720468.2 | XP_006720531.1 | 6 | 519 | |||
XM_011521443.2 | XP_011519745.1 | 5 | 431 | |||
XM_011521444.2 | XP_011519746.1 | 6 | 514 | |||
XM_017022067.1 | XP_016877556.1 | 6 | 519 | |||
GenBank mRNA | AF132733.1 | - | 6 | 519 | ||
AK074870.1 | - | 6 | 512 | |||
AK075403.1 | - | 6 | 549 | |||
AK289591.1 | - | 6 | 512 | |||
AK290484.1 | - | 5 | 446 | |||
AK297816.1 | - | 6 | 508 | |||
AK309915.1 | - | 6 | 512 | |||
AK309954.1 | - | 5 | 452 | |||
BC005335.1 | - | 6 | 515 |