Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSKU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258210.1 | NP_001245139.1 | 2 | 348 | ||
NM_001318477.1 | NP_001305406.1 | 2 | 1245 | |||
NM_001318478.1 | NP_001305407.1 | 2 | 445 | |||
NM_001318479.1 | NP_001305408.1 | 2 | 256 | |||
NM_015516.3 | NP_056331.2 | 2 | 391 | |||
GenBank mRNA | AF191019.1 | - | 1 | 232 | ||
AK075476.1 | - | 2 | 303 | |||
AK126536.1 | - | 2 | 1245 | |||
AK313405.1 | - | 2 | 304 | |||
AY358317.1 | - | 2 | 282 | |||
BC020975.1 | - | 2 | 269 | 3848274 | ||
BT007440.1 | - | 1 | 219 | |||
DA623614.1 | - | 2 | 446 | |||
DA879213.1 | - | 2 | 256 | |||
DB199776.1 | - | 2 | 410 |