Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HINFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243259.1 | NP_001230188.1 | 7 | 980 | ||
NM_015517.4 | NP_056332.2 | 8 | 1097 | |||
NM_198971.2 | NP_945322.1 | 7 | 980 | |||
XM_011542744.2 | XP_011541046.1 | 7 | 1089 | |||
XM_011542745.2 | XP_011541047.1 | 7 | 939 | |||
XM_011542746.1 | XP_011541048.1 | 6 | 760 | |||
XM_017017499.1 | XP_016872988.1 | 7 | 1037 | |||
XM_017017500.1 | XP_016872989.1 | 7 | 916 | |||
XM_017017501.1 | XP_016872990.1 | 8 | 1043 | |||
XM_017017502.1 | XP_016872991.1 | 10 | 1368 | |||
XM_017017503.1 | XP_016872992.1 | 8 | 1133 | |||
XM_017017504.1 | XP_016872993.1 | 8 | 1008 | |||
XM_017017505.1 | XP_016872994.1 | 8 | 986 | |||
XM_017017506.1 | XP_016872995.1 | 8 | 949 | |||
XM_017017507.1 | XP_016872996.1 | 10 | 1188 | |||
XM_017017508.1 | XP_016872997.1 | 9 | 1063 | |||
XM_017017509.1 | XP_016872998.1 | 7 | 815 | |||
GenBank mRNA | AK056362.1 | - | 7 | 893 | ||
AK092869.1 | - | 2 | 1234 | |||
AK301452.1 | - | 7 | 887 | |||
BC001073.2 | - | 7 | 951 | 2820431 | ||
BC001945.1 | - | 7 | 951 | 2820431 | ||
BC012856.1 | - | 7 | 892 | 3871231 | ||
BC017234.2 | - | 7 | 900 | 3537381 |