Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099436.3 | NP_001092906.3 | 3, 4 | 471 | ||
NM_001284364.2 | NP_001271293.2 | 3, 4 | 471 | |||
NM_001284365.2 | NP_001271294.1 | 3, 4 | 394 | |||
NR_104307.2 | 3, 4 | 471 | ||||
NR_104308.2 | 3, 4 | 471 | ||||
NR_104309.2 | 3, 4 | 471 | ||||
XM_005254289.2 | XP_005254346.1 | 4, 5 | 486 | |||
XM_017022068.1 | XP_016877557.1 | 4, 5 | 520 | |||
XM_017022069.1 | XP_016877558.1 | 4, 5 | 524 | |||
XM_017022070.1 | XP_016877559.1 | 4, 5 | 537 | |||
XM_017022071.1 | XP_016877560.1 | 4, 5 | 524 | |||
XM_017022072.1 | XP_016877561.1 | 3, 4 | 700 | |||
GenBank mRNA | AK293181.1 | - | 3, 4 | 324 | ||
AK293650.1 | - | 2, 3 | 489 | |||
AK294241.1 | - | 3, 4 | 405 | |||
AK294245.1 | - | 3, 4 | 401 | |||
AK294251.1 | - | 3, 4 | 405 | |||
AK298879.1 | - | 3, 4 | 328 | |||
AK299380.1 | - | 3, 4 | 398 | |||
AK316102.1 | - | 3, 4 | 405 | |||
AL117482.1 | - | 3, 4 | 606 | |||
BC036117.1 | - | 3, 4 | 442 | |||
BC048289.1 | - | 3, 4 | 399 | |||
BC056423.1 | - | 3, 4 | 389 | |||
BC157884.1 | - | 3, 4 | 453 | 8862706 | ||
BI829133.1 | - | 2, 3 | 555 | |||
BQ927030.1 | - | 3, 4 | 359 |