Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199570.1 | NP_001186499.1 | 10 | 1685 | ||
NM_015526.2 | NP_056341.1 | 11 | 1601 | |||
GenBank mRNA | AJ427922.1 | - | 11 | 1579 | ||
AK090732.1 | - | 11 | 1604 | |||
AK092354.1 | - | 7 | 848 | |||
AK094738.1 | - | 11 | 1599 | |||
AK289509.1 | - | 10 | 1653 | |||
AK294748.1 | - | 11 | 1472 | |||
AL117468.1 | - | 3 | 213 | |||
BC013116.1 | - | 11 | 1513 | 4214441 | ||
BC014486.1 | - | 8 | 1044 | |||
BC128090.1 | - | 7 | 853 |