Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320356.1 | NP_001307285.1 | 7 | 906 | ||
NM_001320357.1 | NP_001307286.1 | 6 | 796 | |||
NM_001320358.1 | NP_001307287.1 | 6 | 666 | |||
NM_001320359.1 | NP_001307288.1 | 7 | 822 | |||
NM_001320360.1 | NP_001307289.1 | 6 | 1052 | |||
NM_001320361.1 | NP_001307290.1 | 7 | 1036 | |||
NM_001320362.1 | NP_001307291.1 | 6 | 616 | |||
NM_001320363.1 | NP_001307292.1 | 6 | 746 | |||
NM_001320364.1 | NP_001307293.1 | 5 | 675 | |||
NM_001320365.1 | NP_001307294.1 | 7 | 986 | |||
NM_015528.2 | NP_056343.1 | 6 | 1182 | |||
XM_017024427.1 | XP_016879916.1 | 5 | 922 | |||
GenBank mRNA | AK025329.1 | - | 6 | 855 | ||
AL050060.1 | - | 4 | 332 | |||
AL537131.3 | - | 7 | 710 | |||
AL834284.1 | - | 5 | 704 | |||
AY203930.1 | - | 5 | 682 | |||
AY960626.1 | - | 5 | 349 | |||
BC010139.2 | - | 5 | 580 | 4128232 | ||
BC013222.1 | - | 5 | 636 | |||
BI825561.1 | - | 6 | 518 | |||
CR457340.1 | - | 5 | 406 |