Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015542.3 | NP_056357.1 | 19 | 3614 | ||
NM_080599.2 | NP_542166.1 | 19 | 3784 | |||
XM_017016034.1 | XP_016871523.1 | 18 | 3270 | |||
GenBank mRNA | AB037829.1 | - | 18 | 3983 | ||
AF301013.1 | - | 19 | 3607 | |||
AF318574.1 | - | 19 | 3584 | |||
AK000764.1 | - | 4 | 445 | |||
AK074375.1 | - | 4 | 377 | |||
AL080198.1 | - | 13 | 1784 | |||
AY013249.1 | - | 18 | 3533 | |||
BC114964.1 | - | 19 | 3578 | 40082739 | ||
BC115737.1 | - | 19 | 3578 | 40082728 |