Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SIPA1L1 SIRNA Assays ›
Gene Symbol:
SIPA1L1
Gene Name:
signal induced proliferation associated 1 like 1
Gene Aliases:
E6TP1
Chromosome Location:
Chr.14: 71320418 - 71741229 on Build GRCh38
UniGene ID:
Hs.654657
Species:
Human
siRNA ID
HSS178018
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SIPA1L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284245.1
NP_001271174.1
5
2199
NM_001284246.1
NP_001271175.1
5
2788
NM_001284247.1
NP_001271176.1
4
2140
NM_015556.2
NP_056371.1
5
2199
XM_005267516.4
XP_005267573.1
5
2788
XM_006720111.2
XP_006720174.1
7
4078
XM_011536630.1
XP_011534932.1
9
2550
XM_011536631.1
XP_011534933.1
9
2961
XM_011536632.1
XP_011534934.1
8
2470
XM_011536635.2
XP_011534937.1
8
2617
XM_011536638.2
XP_011534940.1
4
1273
XM_017021159.1
XP_016876648.1
11
2707
XM_017021160.1
XP_016876649.1
9
2629
XM_017021161.1
XP_016876650.1
10
2582
XM_017021162.1
XP_016876651.1
12
2764
XM_017021163.1
XP_016876652.1
7
2431
XM_017021164.1
XP_016876653.1
7
2438
XM_017021165.1
XP_016876654.1
9
2507
XM_017021166.1
XP_016876655.1
10
2640
XM_017021167.1
XP_016876656.1
9
2515
XM_017021168.1
XP_016876657.1
9
2489
XM_017021169.1
XP_016876658.1
8
2642
XM_017021170.1
XP_016876659.1
8
3000
XM_017021171.1
XP_016876660.1
9
2573
XM_017021172.1
XP_016876661.1
8
2448
XM_017021173.1
XP_016876662.1
7
2575
XM_017021174.1
XP_016876663.1
7
2815
XM_017021175.1
XP_016876664.1
7
2933
XM_017021176.1
XP_016876665.1
12
2760
XM_017021177.1
XP_016876666.1
12
2749
XM_017021178.1
XP_016876667.1
10
2587
XM_017021179.1
XP_016876668.1
9
2490
XM_017021180.1
XP_016876669.1
13
2802
XM_017021181.1
XP_016876670.1
11
2707
XM_017021182.1
XP_016876671.1
10
2640
XM_017021183.1
XP_016876672.1
8
2448
XM_017021184.1
XP_016876673.1
12
2760
XM_017021185.1
XP_016876674.1
5
2788
XM_017021186.1
XP_016876675.1
11
2707
XM_017021187.1
XP_016876676.1
9
2573
XM_017021188.1
XP_016876677.1
8
2448
XM_017021189.1
XP_016876678.1
10
2557
XM_017021190.1
XP_016876679.1
11
2707
XM_017021191.1
XP_016876680.1
9
2515
XM_017021192.1
XP_016876681.1
8
2448
XM_017021193.1
XP_016876682.1
8
2446
XM_017021194.1
XP_016876683.1
11
2707
XM_017021195.1
XP_016876684.1
11
2707
XM_017021196.1
XP_016876685.1
11
2707
XM_017021197.1
XP_016876686.1
10
907
XM_017021198.1
XP_016876687.1
7
648
XM_017021199.1
XP_016876688.1
11
949
XM_017021200.1
XP_016876689.1
8
715
GenBank mRNA
AB007900.2
-
4
1923
AF090989.1
-
5
2186
AF090990.1
-
5
2186
AK122930.1
-
5
2788
AK302650.1
-
4
438
BC146777.1
-
4
1923
9007226
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
actin cytoskeleton reorganization
regulation of GTPase activity
ephrin receptor signaling pathway
regulation of synaptic plasticity
regulation of axonogenesis
regulation of small GTPase mediated signal transduction
regulation of dendritic spine morphogenesis
activation of GTPase activity
molecular_function
GTPase activator activity
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline