Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015557.2 | NP_056372.1 | 40 | 5924 | ||
GenBank mRNA | AB007913.1 | - | 22 | 2885 | ||
AF425231.1 | - | 40 | 5913 | |||
AK127046.1 | - | 32 | 4531 | |||
AL117491.1 | - | 30 | 4117 |