Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130110.1 | NP_001123582.1 | 2 | 499 | ||
NM_015559.2 | NP_056374.2 | 2 | 400 | |||
GenBank mRNA | AB007897.2 | - | 2 | 400 | ||
BC062338.1 | - | 2 | 499 | |||
BC146776.1 | - | 2 | 400 | 9007225 |