Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142782.1 | NP_001136254.1 | 10 | 1627 | ||
NM_152900.2 | NP_690864.2 | 10 | 1627 | |||
XM_005270737.3 | XP_005270794.1 | 10 | 1627 | |||
XM_011541208.2 | XP_011539510.1 | 10 | 3578 | |||
XM_017000974.1 | XP_016856463.1 | 10 | 2006 | |||
XM_017000975.1 | XP_016856464.1 | 4 | 599 | |||
GenBank mRNA | AB046854.1 | - | 6 | 693 | ||
AF213259.1 | - | 10 | 1446 | |||
BC130409.1 | - | 10 | 1627 | 40146440 |