Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015470.2 | NP_056285.1 | 3 | 1762 | ||
XM_005264251.3 | XP_005264308.1 | 3 | 1703 | |||
XM_005264252.3 | XP_005264309.1 | 3 | 1703 | |||
XM_005264253.4 | XP_005264310.1 | 3 | 1703 | |||
XM_006711985.3 | XP_006712048.1 | 3 | 1703 | |||
XM_011532753.2 | XP_011531055.1 | 3 | 1213 | |||
XM_011532754.2 | XP_011531056.1 | 3 | 1703 | |||
XM_011532755.2 | XP_011531057.1 | 3 | 1703 | |||
XM_011532756.2 | XP_011531058.1 | 3 | 1703 | |||
XM_017003788.1 | XP_016859277.1 | 3 | 1265 | |||
GenBank mRNA | AB020664.1 | - | 3 | 1762 | ||
AF153085.1 | - | 3 | 1210 | |||
AF334812.1 | - | 3 | 1761 | |||
AK023823.1 | - | 2 | 1427 | |||
BC035013.1 | - | 3 | 1685 | 5269818 |