Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278185.1 | NP_001265114.1 | 22 | 2779 | ||
NM_001278186.1 | NP_001265115.1 | 21 | 2737 | |||
NM_015601.3 | NP_056416.2 | 23 | 2989 | |||
NM_022079.2 | NP_071362.1 | 24 | 3013 | |||
XM_011539592.2 | XP_011537894.1 | 25 | 3087 | |||
XM_011539593.2 | XP_011537895.1 | 24 | 3063 | |||
XM_011539594.2 | XP_011537896.1 | 26 | 3173 | |||
XM_011539595.2 | XP_011537897.1 | 26 | 3223 | |||
XM_011539596.2 | XP_011537898.1 | 25 | 3101 | |||
XM_011539597.2 | XP_011537899.1 | 23 | 2724 | |||
XM_017016041.1 | XP_016871530.1 | 22 | 2652 | |||
GenBank mRNA | AB046813.1 | - | 21 | 2867 | ||
AK027835.1 | - | 14 | 1552 | |||
AK292809.1 | - | 23 | 2869 | |||
AY221963.1 | - | 24 | 2950 | |||
AY650032.1 | - | 21 | 2674 | |||
AY650033.1 | - | 22 | 2716 | |||
BC015043.1 | - | 21 | 2640 | |||
BC039600.1 | - | 23 | 2899 | 5212048 | ||
BX537563.1 | - | 23 | 5476 |