Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015621.2 | NP_056436.2 | 7 | 685 | ||
GenBank mRNA | AK023767.1 | - | 6 | 463 | ||
AK225826.1 | - | 7 | 655 | |||
AK292841.1 | - | 7 | 685 | |||
AL713742.1 | - | 6 | 609 | |||
BC016647.1 | - | 6 | 523 | |||
BM927471.1 | - | 2 | 202 |