Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015662.2 | NP_056477.1 | 30 | 3470 | ||
XM_005264254.1 | XP_005264311.1 | 30 | 3301 | |||
XM_006711986.3 | XP_006712049.1 | 31 | 3562 | |||
XM_006711987.1 | XP_006712050.1 | 30 | 3367 | |||
XM_011532757.2 | XP_011531059.1 | 30 | 3676 | |||
XM_011532758.1 | XP_011531060.1 | 30 | 3367 | |||
XM_011532759.2 | XP_011531061.1 | 16 | 1952 | |||
XM_011532760.2 | XP_011531062.1 | 13 | 1632 | |||
XM_017003790.1 | XP_016859279.1 | 31 | 3496 | |||
XM_017003791.1 | XP_016859280.1 | 29 | 3437 | |||
XM_017003792.1 | XP_016859281.1 | 30 | 3367 | |||
XM_017003793.1 | XP_016859282.1 | 26 | 2877 | |||
XM_017003794.1 | XP_016859283.1 | 24 | 2580 | |||
XM_017003795.1 | XP_016859284.1 | 28 | 3429 | |||
GenBank mRNA | AB033005.1 | - | 12 | 1335 | ||
AL110218.1 | - | 27 | 3026 | |||
BC137126.1 | - | 30 | 3360 | 9021123 |