Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012158.2 | NP_036290.1 | 5 | 1063 | ||
XM_005266336.1 | XP_005266393.1 | 5 | 846 | |||
XM_005266337.2 | XP_005266394.1 | 4 | 698 | |||
GenBank mRNA | AF126028.1 | - | 4 | 667 | ||
AF129532.1 | - | 5 | 730 | |||
AK025484.1 | - | 3 | 261 | |||
AK314442.1 | - | 5 | 1031 | |||
AL833187.1 | - | 5 | 1011 | |||
BC072448.1 | - | 5 | 1017 | 6166244 |