Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037174.1 | NP_001032251.1 | 3 | 213 | ||
NM_012097.3 | NP_036229.1 | 3 | 469 | |||
NM_177985.2 | NP_817114.2 | 3 | 211 | |||
GenBank mRNA | AF087889.1 | - | 399 | |||
AF100740.1 | - | 3 | 216 | |||
AF493891.1 | - | 3 | 150 | |||
AK292714.1 | - | 3 | 379 | |||
AK298937.1 | - | 3 | 157 | |||
BC001254.1 | - | 3 | 387 | 3453177 | ||
BG707385.1 | - | 3 | 190 | |||
BQ216648.1 | - | 3 | 456 | |||
BT009829.1 | - | 3 | 150 | |||
BX461356.2 | - | 3 | 184 | |||
CX867116.1 | - | 3 | 475 |