Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004055.1 | NP_001004055.1 | 5 | 655 | ||
NM_012163.2 | NP_036295.1 | 5 | 820 | |||
XM_017023126.1 | XP_016878615.1 | 5 | 1084 | |||
XM_017023127.1 | XP_016878616.1 | 5 | 1335 | |||
XM_017023128.1 | XP_016878617.1 | 5 | 1140 | |||
XM_017023129.1 | XP_016878618.1 | 5 | 2226 | |||
XM_017023130.1 | XP_016878619.1 | 5 | 808 | |||
XM_017023131.1 | XP_016878620.1 | 5 | 667 | |||
XM_017023132.1 | XP_016878621.1 | 5 | 970 | |||
XM_017023133.1 | XP_016878622.1 | 5 | 996 | |||
XM_017023134.1 | XP_016878623.1 | 5 | 775 | |||
GenBank mRNA | AF176701.1 | - | 5 | 665 | ||
AK096101.1 | - | 3 | 1292 | |||
AK316602.1 | - | 5 | 668 | |||
BC018785.1 | - | 5 | 820 | 4875646 | ||
BI085522.1 | - | 4 | 537 |