Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012168.5 | NP_036300.2 | 5, 6 | 1246 | ||
GenBank mRNA | AF174594.1 | - | 4, 5 | 638 | ||
AF187318.1 | - | 5, 6 | 1094 | |||
AK313019.1 | - | 5, 6 | 846 | |||
AW138262.1 | - | 1, 2 | 451 | |||
AW162121.1 | - | 1, 2 | 445 | |||
BC025233.1 | - | 5, 6 | 830 | 5090925 | ||
BC096747.1 | - | 5, 6 | 846 | 5259298 | ||
CR542110.1 | - | 5, 6 | 755 |