Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202469.1 | NP_001189398.1 | 8 | 744 | ||
NM_001483.2 | NP_001474.1 | 10 | 861 | |||
GenBank mRNA | AF029786.1 | - | 10 | 836 | ||
AJ001259.1 | - | 10 | 828 | |||
AK097049.1 | - | 8 | 730 | |||
AK125036.1 | - | 7 | 3117 | |||
AK315648.1 | - | 10 | 846 | |||
BC000732.2 | - | 10 | 834 | 2959662 | ||
BC001837.2 | - | 10 | 834 | 2959662 | ||
BC030821.1 | - | 10 | 843 | 4808962 | ||
BT007112.1 | - | 10 | 828 | |||
CR407612.1 | - | 10 | 828 |