Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014365.2 | NP_055180.1 | 2 | 915 | ||
GenBank mRNA | AF133207.1 | - | 2 | 752 | ||
AF191017.1 | - | 2 | 915 | |||
AF217987.1 | - | 2 | 760 | |||
AF250138.1 | - | 2 | 400 | |||
AK312501.1 | - | 2 | 768 | |||
AL136936.1 | - | 2 | 745 | |||
BC002673.2 | - | 2 | 883 | 3607117 | ||
BT006876.1 | - | 2 | 392 | |||
CR533453.1 | - | 2 | 392 |