Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021078.2 | NP_066564.2 | 5 | 917 | ||
XM_006721817.3 | XP_006721880.1 | 5 | 946 | |||
XM_006721818.3 | XP_006721881.1 | 5 | 638 | |||
XM_017024448.1 | XP_016879937.1 | 5 | 889 | |||
GenBank mRNA | AF029777.1 | - | 5 | 881 | ||
BC032743.1 | - | 5 | 894 | 5575574 | ||
BC039907.1 | - | 5 | 909 | 6064838 | ||
BC105977.1 | - | 5 | 886 | 6137532 |