Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NARF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038618.2 | NP_001033707.1 | 6 | 908 | ||
NM_001083608.1 | NP_001077077.1 | 5 | 606 | |||
NM_012336.3 | NP_036468.1 | 6 | 750 | |||
NM_031968.2 | NP_114174.1 | 6 | 750 | |||
XM_005256340.3 | XP_005256397.1 | 6 | 916 | |||
XM_006722278.3 | XP_006722341.1 | 5 | 703 | |||
XM_011523541.2 | XP_011521843.1 | 6 | 840 | |||
XM_011523543.2 | XP_011521845.1 | 7 | 753 | |||
XM_011523544.2 | XP_011521846.1 | 6 | 847 | |||
XM_017024449.1 | XP_016879938.1 | 6 | 893 | |||
XM_017024450.1 | XP_016879939.1 | 7 | 990 | |||
GenBank mRNA | AF128406.1 | - | 6 | 701 | ||
AK000929.1 | - | 6 | 703 | |||
AK056966.1 | - | 6 | 883 | |||
AK297875.1 | - | 5 | 615 | |||
AK303155.1 | - | 6 | 665 | |||
AL137729.1 | - | 3 | 341 | |||
BC000438.2 | - | 6 | 717 | 2820621 | ||
BC016440.1 | - | 5 | 493 | 3447710 |