Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012434.4 | NP_036566.1 | 11 | 1512 | ||
XM_005248710.3 | XP_005248767.1 | 11 | 1534 | |||
XM_005248711.2 | XP_005248768.1 | 11 | 1766 | |||
XM_011535750.2 | XP_011534052.1 | 10 | 1322 | |||
XM_017010730.1 | XP_016866219.1 | 10 | 1678 | |||
XM_017010731.1 | XP_016866220.1 | 10 | 1528 | |||
GenBank mRNA | AF244577.1 | - | 12 | 1769 | ||
AJ387747.1 | - | 11 | 1654 | |||
AK025880.1 | - | 5 | 425 | |||
AK026921.1 | - | 3 | 177 | |||
AK075320.1 | - | 12 | 1568 | |||
AK315789.1 | - | 11 | 1470 | |||
BC020961.2 | - | 11 | 1471 | 3847279 |