Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282509.1 | NP_001269438.1 | 2 | 388 | ||
NM_001282510.1 | NP_001269439.1 | 2 | 388 | |||
NM_012466.3 | NP_036598.1 | 2 | 388 | |||
NR_104199.1 | 2 | 388 | ||||
XM_011527901.1 | XP_011526203.1 | 2 | 383 | |||
XM_011527902.1 | XP_011526204.1 | 2 | 383 | |||
XM_011527903.2 | XP_011526205.1 | 2 | 319 | |||
GenBank mRNA | BC029908.1 | - | 2 | 358 | 5168677 | |
BM560276.1 | - | 2 | 375 | |||
DB457828.1 | - | 2 | 378 | |||
HY051653.1 | - | 2 | 389 |