Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018959.3 | NP_061832.2 | 7 | 706 | ||
NM_170711.2 | NP_733829.1 | 7 | 706 | |||
XM_005259531.3 | XP_005259588.1 | 7 | 656 | |||
XM_011527904.2 | XP_011526206.1 | 7 | 1971 | |||
XM_011527905.2 | XP_011526207.1 | 7 | 1979 | |||
XM_011527906.2 | XP_011526208.1 | 7 | 1976 | |||
XM_011527907.2 | XP_011526209.1 | 7 | 1985 | |||
XM_011527908.2 | XP_011526210.1 | 7 | 1978 | |||
XM_011527909.2 | XP_011526211.1 | 7 | 1978 | |||
XM_011527910.2 | XP_011526212.1 | 7 | 1058 | |||
XM_017026581.1 | XP_016882070.1 | 7 | 701 | |||
GenBank mRNA | AF181719.1 | - | 7 | 609 | ||
AK056850.1 | - | 5 | 329 | |||
BC012062.1 | - | 7 | 574 | 4549444 | ||
BI909277.1 | - | 7 | 690 | |||
KU178593.1 | - | 7 | 501 |