Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153487.3 | NP_705691.1 | 1 | 1191 | ||
XM_006715056.3 | XP_006715119.1 | 1 | 135 | |||
XM_017010734.1 | XP_016866223.1 | 1 | 135 | |||
GenBank mRNA | AF478693.1 | - | 1 | 1191 |