Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153615.1 | NP_705843.1 | 6 | 2241 | ||
GenBank mRNA | AK131340.1 | - | 8 | 2679 | ||
AY101396.1 | - | 6 | 2241 | |||
BC101108.3 | - | 5 | 1921 | 40011557 | ||
BC101109.2 | - | 5 | 1921 | 40011564 | ||
BC101110.3 | - | 6 | 1796 |
Gene Symbol: GUSBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024448.2 | 8 | 1672 | |||
GenBank mRNA | AK056875.1 | - | 8 | 1602 | ||
AK091837.1 | - | 7 | 1410 | |||
BX538181.1 | - | 7 | 1397 |