Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005261.3 | NP_005252.1 | 2 | 510 | ||
NM_181702.2 | NP_859053.1 | 2 | 395 | |||
XM_017013315.1 | XP_016868804.1 | 2 | 536 | |||
GenBank mRNA | AK314017.1 | - | 2 | 395 | ||
BC022010.2 | - | 2 | 395 | 4823985 | ||
BI759640.1 | - | 2 | 405 | |||
DB018783.1 | - | 2 | 510 | |||
U10550.1 | - | 2 | 484 | |||
U13052.1 | - | 1 | 271 |