Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012345.2 | NP_036477.2 | 9 | 1299 | ||
GenBank mRNA | AF159548.1 | - | 9 | 1257 | ||
AK314242.1 | - | 9 | 1292 | |||
BC017745.1 | - | 7 | 782 | 4398678 |