Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173393.2 | NP_001166864.1 | 2, 3 | 251 | ||
NM_001308156.1 | NP_001295085.1 | 2, 3 | 372 | |||
NM_012206.3 | NP_036338.2 | 1, 2 | 338 | |||
XM_017009339.1 | XP_016864828.1 | 1, 2 | 52 | |||
XM_017009340.1 | XP_016864829.1 | 1, 2 | 52 | |||
GenBank mRNA | AF043724.1 | - | 2, 3 | 91 | ||
AW604297.1 | - | 2, 3 | 381 | |||
BC013325.1 | - | 1, 2 | 208 | 3826378 | ||
BG752287.1 | - | 1, 2 | 554 | |||
CR457114.1 | - | 1, 2 | 40 | |||
DA384612.1 | - | 2, 3 | 95 | |||
HQ412639.1 | - | 1, 2 | 40 | |||
HQ412640.1 | - | 1, 2 | 40 | |||
HQ412641.1 | - | 1, 2 | 40 | |||
JX049978.1 | - | 1, 2 | 40 |
Gene Symbol: HAVCR1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003603.1 | 1 | 53 | |||
GenBank mRNA | BX114328.1 | - | 1 | 659 |