Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005142.2 | NP_005133.2 | 5 | 645 | ||
XM_011544939.2 | XP_011543241.1 | 5 | 673 | |||
GenBank mRNA | AK301295.1 | - | 5 | 757 | ||
AK314275.1 | - | 5 | 664 | |||
BC037958.1 | - | 5 | 645 | 5759934 | ||
X76562.1 | - | 5 | 644 |