Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012133.5 | NP_036265.3 | 20, 21 | 2240 | ||
GenBank mRNA | AB047847.1 | - | 20, 21 | 2150 | ||
AF157833.1 | - | 20, 21 | 2210 | |||
AF207598.1 | - | 20, 21 | 2206 | |||
AF308295.1 | - | 1, 2 | 103 | |||
AK002086.1 | - | 12, 13 | 1542 | |||
AK022934.1 | - | 20, 21 | 2137 | |||
AK024414.1 | - | 13, 14 | 1618 | |||
AK291409.1 | - | 20, 21 | 2230 | |||
BC017443.1 | - | 20, 21 | 2133 |