Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHORDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144073.1 | NP_001137545.1 | 10 | 1302 | ||
NM_012124.2 | NP_036256.2 | 11 | 1359 | |||
XM_017017540.1 | XP_016873029.1 | 12 | 1137 | |||
XM_017017541.1 | XP_016873030.1 | 11 | 1073 | |||
GenBank mRNA | AF123249.1 | - | 10 | 997 | ||
AF192466.1 | - | 11 | 1033 | |||
AF217505.1 | - | 6 | 701 | |||
AK225365.1 | - | 11 | 1061 | |||
AK225565.1 | - | 11 | 1044 | |||
AK226099.1 | - | 9 | 4361 | |||
AK290231.1 | - | 11 | 1359 | |||
AK312663.1 | - | 11 | 1044 | |||
BC017789.1 | - | 4 | 388 | |||
BC072461.1 | - | 11 | 1008 | 6471725 |
Gene Symbol: LOC727896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026659.1 | 1 | 1028 |