Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271815.1 | NP_001258744.1 | 7 | 931 | ||
NM_007052.4 | NP_008983.2 | 8 | 1042 | |||
NM_013955.2 | NP_039249.1 | 8 | 1042 | |||
XM_017029407.1 | XP_016884896.1 | 6 | 691 | |||
GenBank mRNA | AF127763.2 | - | 1042 | |||
AF166327.1 | - | 8 | 906 | |||
AF166328.1 | - | 8 | 906 | |||
AK292201.1 | - | 8 | 997 | |||
AK313678.1 | - | 8 | 951 | |||
BC075014.2 | - | 8 | 857 | 30915618 | ||
BC075015.2 | - | 8 | 857 | 30915405 | ||
DQ424904.1 | - | 7 | 725 |