Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014987.1 | NP_001014987.1 | 1, 2 | 450 | ||
NM_001014988.1 | NP_001014988.1 | 1, 2 | 450 | |||
NM_001014989.1 | NP_001014989.2 | 2, 3 | 242 | |||
NM_014387.3 | NP_055202.1 | 1, 2 | 450 | |||
GenBank mRNA | AF036905.1 | - | 1, 2 | 155 | ||
AF036906.1 | - | 1, 2 | 176 | |||
AJ223280.1 | - | 1, 2 | 420 | |||
AK307342.1 | - | 1, 2 | 467 | |||
BC011563.1 | - | 2, 3 | 242 | 4644259 | ||
BI769296.1 | - | 1, 2 | 451 | |||
BI911769.1 | - | 1, 2 | 417 | |||
FN432831.1 | - | 1, 2 | 98 | |||
FN432832.1 | - | 1, 2 | 98 | |||
FN432833.1 | - | 1, 2 | 98 | |||
FN432834.1 | - | 1, 2 | 98 |