Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014421.2 | NP_055236.1 | 3 | 1218 | ||
GenBank mRNA | AB033208.1 | - | 3 | 735 | ||
AB033941.1 | - | 3 | 735 | |||
AF177395.1 | - | 3 | 1230 | |||
AK312695.1 | - | 3 | 744 | |||
AY358414.1 | - | 3 | 723 | |||
BC075077.2 | - | 3 | 547 | 30915382 | ||
BC075078.2 | - | 3 | 547 | 30915595 | ||
BC126328.1 | - | 3 | 819 | 8992044 | ||
BC126330.1 | - | 3 | 819 | 8992046 |