Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014433.2 | NP_055248.1 | 2, 3 | 368 | ||
XM_011530149.2 | XP_011528451.1 | 2, 3 | 407 | |||
XM_011530150.1 | XP_011528452.1 | 2, 3 | 407 | |||
XM_011530151.2 | XP_011528453.1 | 2, 3 | 570 | |||
XM_011530152.2 | XP_011528454.1 | 2, 3 | 498 | |||
XM_011530154.2 | XP_011528456.1 | 1, 2 | 113 | |||
XM_017028774.1 | XP_016884263.1 | 2, 3 | 393 | |||
GenBank mRNA | AF133587.1 | - | 2, 3 | 368 | ||
AK299530.1 | - | 2, 3 | 326 | |||
AK310327.1 | - | 2, 3 | 407 | |||
BC008986.1 | - | 2, 3 | 336 | 4339021 | ||
CR456567.1 | - | 1, 2 | 268 |