Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164617.1 | NP_001158089.1 | 5 | 1338 | ||
NM_001164618.1 | NP_001158090.1 | 4 | 1221 | |||
NM_001164619.1 | NP_001158091.1 | 3 | 1107 | |||
NM_004484.3 | NP_004475.1 | 4 | 1269 | |||
XM_017029413.1 | XP_016884902.1 | 4 | 1240 | |||
GenBank mRNA | AK222761.1 | - | 4 | 1217 | ||
AK222766.1 | - | 4 | 1217 | |||
AK300168.1 | - | 4 | 1192 | |||
AK310196.1 | - | 3 | 936 | |||
AK310689.1 | - | 5 | 1288 | |||
DQ349136.1 | - | 3 | 910 | |||
DQ349137.1 | - | 4 | 1072 | |||
DQ349138.1 | - | 5 | 1141 | |||
JQ943686.1 | - | 4 | 1072 | |||
L47125.1 | - | 4 | 1223 | |||
L47176.1 | - | 4 | 1218 | |||
U50410.1 | - | 4 | 1216 | |||
Z37987.1 | - | 4 | 1180 |