Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000404.3 | NP_000395.2 | 5 | 673 | ||
NM_001079811.2 | NP_001073279.1 | 5 | 567 | |||
NM_001135602.2 | NP_001129074.1 | 3 | 461 | |||
NM_001317040.1 | NP_001303969.1 | 6 | 817 | |||
GenBank mRNA | AK130133.1 | - | 5 | 601 | ||
AK222763.1 | - | 5 | 564 | |||
AK223091.1 | - | 5 | 589 | |||
AK298985.1 | - | 3 | 373 | |||
AK300021.1 | - | 5 | 500 | |||
AK300753.1 | - | 6 | 733 | |||
AK312988.1 | - | 5 | 560 | |||
BC007493.2 | - | 5 | 575 | 2988086 | ||
BT007147.1 | - | 5 | 528 | |||
M22590.1 | - | 5 | 562 | |||
M27507.1 | - | 5 | 578 | |||
M27508.1 | - | 3 | 366 | |||
M34423.1 | - | 5 | 588 |