Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPKOW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015698.5 | NP_056513.2 | 11 | 1467 | ||
XM_017029415.1 | XP_016884904.1 | 12 | 1541 | |||
GenBank mRNA | AB209806.1 | - | 11 | 1387 | ||
BC000397.2 | - | 11 | 1395 | 2822421 | ||
BC003148.1 | - | 11 | 1395 | 2822421 | ||
U66359.1 | - | 11 | 1444 |