Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SESN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199933.1 | NP_001186862.1 | 4 | 497 | ||
NM_001199934.1 | NP_001186863.1 | 4 | 539 | |||
NM_014454.2 | NP_055269.1 | 4 | 1011 | |||
GenBank mRNA | AF033120.1 | - | 4 | 490 | ||
AF033121.1 | - | 4 | 539 | |||
AF033122.1 | - | 4 | 590 | |||
AK290157.1 | - | 4 | 818 | |||
AK290551.1 | - | 4 | 494 | |||
BC112036.1 | - | 4 | 643 | 8327504 | ||
BC113569.1 | - | 4 | 631 | 8322621 | ||
BI549226.1 | - | 4 | 501 |